Skip to Content
Merck
All Photos(1)

Key Documents

183474

Sigma-Aldrich

2-Nitrophenethyl alcohol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$164.00

$164.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$164.00

About This Item

Linear Formula:
O2NC6H4CH2CH2OH
CAS Number:
Molecular Weight:
167.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$164.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.5637 (lit.)

bp

267 °C (lit.)

mp

2 °C (lit.)

density

1.19 g/mL at 25 °C (lit.)

SMILES string

OCCc1ccccc1[N+]([O-])=O

InChI

1S/C8H9NO3/c10-6-5-7-3-1-2-4-8(7)9(11)12/h1-4,10H,5-6H2

InChI key

SLRIOXRBAPBGEI-UHFFFAOYSA-N

Application

2-Nitrophenethyl alcohol was used in the synthesis of indoles[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruthenium-catalyzed dehydrogenative N-heterocyclization. Indoles from 2-aminophenethyl alcohols and 2-nitrophenethyl alcohols.
Tsuji Y, et al.
The Journal of Organic Chemistry, 55(2), 580-584 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service