Skip to Content
Merck
All Photos(1)

Documents

183113

Sigma-Aldrich

3,3-Dimethylbutylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2CH2NH2
CAS Number:
Molecular Weight:
101.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.4135 (lit.)

bp

114-116 °C (lit.)

density

0.752 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)CCN

InChI

1S/C6H15N/c1-6(2,3)4-5-7/h4-5,7H2,1-3H3

InChI key

GPWHFPWZAPOYNO-UHFFFAOYSA-N

General description

3,3-Dimethylbutylamine forms a monoclinic crystal with cyclomaltoheptaose.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

I M Mavridis et al.
Carbohydrate research, 220, 11-21 (1991-11-11)
The crystal of the 1:1 complex of 3,3-dimethylbutylamine with cyclomaltoheptaose (beta CD, beta-cyclodextrin) is monoclinic C2 with a = 19.187(9), b = 24.56(1), c = 15.893(7) A, beta = 108.77(4) degrees, V = 7091 A3, and Z = 4. Two
Pieter A Cohen et al.
Clinical toxicology (Philadelphia, Pa.), 56(6), 421-426 (2017-11-09)
The United States Food and Drug Administration banned the stimulant 1,3-dimethylamylamine (1,3-DMAA) from dietary supplements and warned consumers that the stimulant can pose cardiovascular risks ranging from high blood pressure to heart attacks. We designed our study to determine if

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service