Skip to Content
Merck
All Photos(1)

Documents

182656

Sigma-Aldrich

Poly(ethyl vinyl ether)

average Mw ~3,800 by GPC

Synonym(s):

Ethyl vinyl ether homopolymer, Vinyl ethyl ether polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH(OC2H5)-]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

22.5 mmHg ( 20 °C)

autoignition temp.

446 °F

mol wt

average Mw ~3,800 by GPC

refractive index

n20/D 1.454 (lit.)

transition temp

Tg −60 °C

density

0.968 g/mL at 25 °C (lit.)

SMILES string

CCOC=C

InChI

1S/C4H8O/c1-3-5-4-2/h3H,1,4H2,2H3

InChI key

FJKIXWOMBXYWOQ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service