Skip to Content
Merck
All Photos(1)

Key Documents

181900

Sigma-Aldrich

Polyethylene

High density, average Mw ~125,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(CH2CH2)nH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

beads

mol wt

average Mw ~125,000

transition temp

Tm 130 °C

density

0.95 g/mL at 25 °C

InChI

1S/C2H4/c1-2/h1-2H2

InChI key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Frederick J Kummer et al.
Journal of long-term effects of medical implants, 22(2), 177-179 (2013-02-23)
A polyethylene cup explanted after 41 years was examined using several analytical techniques to determine whether there was a material cause for the extremely low wear observed. Neither the amount of polyethylene oxidation nor crystallinity appeared to be a factor.
Lisa K McCarthy et al.
Pediatrics, 132(1), e135-e141 (2013-06-19)
Hypothermia on admission to the NICU is associated with increased mortality in preterm infants. Many newborns are hypothermic on admission despite using polyethylene bags (PBs). Using exothermic mattresses (EMs) in addition to PBs may reduce hypothermia but increase hyperthermia. We
Katharine S Woods et al.
American journal of veterinary research, 74(4), 523-529 (2013-03-28)
To compare the diagnostic quality of bronchoalveolar lavage (BAL) fluid acquired from healthy dogs by manual aspiration via polyethylene tubing (MAPT) and via suction pump aspiration (SPA) with a suction trap connection. 12 healthy adult Beagles. BAL was performed with
Agnieszka Karbownik et al.
Acta poloniae pharmaceutica, 70(2), 301-307 (2013-04-26)
The study concerned the stability of calcium folinate in concentrate in glass vials and diluted in polyethylene (PE) bags stored at 15-25 degrees C and 2-8 degrees C for up to 34 days. Original vials of calcium folinate injection (10
Nina Bloecher et al.
Biofouling, 29(3), 237-246 (2013-02-27)
The hydroid Ectopleura larynx is one of the main fouling organisms on salmon aquaculture cages in Norway; this study investigated novel surface materials and microtopographies to deter its settlement. The settlement preferences of hydroid larvae for 12 materials with wettabilities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service