Skip to Content
Merck
All Photos(2)

Key Documents

180696

Sigma-Aldrich

Dimethyl 2-oxopropylphosphonate

95%

Synonym(s):

Dimethyl acetonylphosphonate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$49.20
25 G
$234.00

$49.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$49.20
25 G
$234.00

About This Item

Linear Formula:
CH3COCH2P(O)(OCH3)2
CAS Number:
Molecular Weight:
166.11
Beilstein:
1766218
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$49.20


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.439 (lit.)

bp

76-79 °C/3 mmHg (lit.)

density

1.202 g/mL at 25 °C (lit.)

functional group

ketone
phosphonate

SMILES string

COP(=O)(CC(C)=O)OC

InChI

1S/C5H11O4P/c1-5(6)4-10(7,8-2)9-3/h4H2,1-3H3

InChI key

UOWIYNWMROWVDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used together with tosyl azide in a one-pot alkynylation of aldehydes.[1][2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 753-753 (2006)
Synthesis, 59-59 (2004)
B Resul et al.
Journal of medicinal chemistry, 36(2), 243-248 (1993-01-22)
A series of phenyl-substituted analogues of prostaglandin F2 alpha (PGF2 alpha) were prepared and evaluated for ocular hypotensive effect and side effects in different animal models. In addition, the activity of the analogues on FP receptors was studied in vitro.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service