Skip to Content
Merck
All Photos(2)

Key Documents

177199

Sigma-Aldrich

Methyl propargyl ether

≥97%

Synonym(s):

3-Methoxy-1-propyne, Methyl 2-propynyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$248.00
50 G
$1,070.00

$248.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$248.00
50 G
$1,070.00

About This Item

Linear Formula:
HC≡CCH2OCH3
CAS Number:
Molecular Weight:
70.09
Beilstein:
878166
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$248.00


Check Cart for Availability

Request a Bulk Order

Assay

≥97%

form

liquid

refractive index

n20/D 1.396 (lit.)

bp

61-62 °C (lit.)

density

0.83 g/mL at 25 °C (lit.)

functional group

ether

storage temp.

2-8°C

SMILES string

COCC#C

InChI

1S/C4H6O/c1-3-4-5-2/h1H,4H2,2H3

InChI key

YACFFSVYSPMSGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The Raman spectra of methyl propargyl ether has been studied[1].

Application

Methyl propargyl ether was used in the synthesis of [Ru3(μ3-κ(2)-HNNMe2)(μ3-κ(2)-MeOCH2CCH2)(μ-CO)2(CO)6][2].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javier A Cabeza et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 11(20), 6040-6052 (2005-07-30)
The reactions of the hydrido-triruthenium cluster complex [Ru3(mu-H)(mu3-kappa(2)-HNNMe2)(CO)9] (1; H2NNMe2 = 1,1-dimethylhydrazine) with alkynes that have alpha-hydrogen atoms give trinuclear derivatives containing edge-bridging allyl or face-capping alkenyl ligands. Under mild conditions (THF, 70 degrees C) the isolated products are as
Conformational stability, barriers to internal rotation, ab initio calculations and vibrational assignment of methyl propargyl ether.
Journal of Molecular Structure, 320, 193-216 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service