Skip to Content
Merck
All Photos(1)

Key Documents

177148

Sigma-Aldrich

3-Quinolinecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$55.20
5 G
$158.00
25 G
$296.00

$55.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$55.20
5 G
$158.00
25 G
$296.00

About This Item

Empirical Formula (Hill Notation):
C10H7NO2
CAS Number:
Molecular Weight:
173.17
Beilstein:
126542
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$55.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

277-280 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1cnc2ccccc2c1

InChI

1S/C10H7NO2/c12-10(13)8-5-7-3-1-2-4-9(7)11-6-8/h1-6H,(H,12,13)

InChI key

DJXNJVFEFSWHLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The antibacterial activity of 3-quinolinecarboxylic acid derivatives were evaluated[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M P Wentland et al.
Journal of medicinal chemistry, 27(9), 1103-1108 (1984-09-01)
A series of novel 3-quinolinecarboxylic acid derivatives have been prepared and their antibacterial activity evaluated. These derivatives are characterized by fluorine attached to the 6-position and substituted amino groups appended to the 1- and 7-positions. Structure-activity relationship studies indicate that
Antonello Mai et al.
Journal of medicinal chemistry, 49(23), 6897-6907 (2006-12-13)
Starting from a yeast phenotypic screening performed on 21 compounds, we described the identification of two small molecules (9 and 18) able to significantly reduce the S. cerevisiae cell growth, thus miming the effect of GCN5 deletion mutant. Tested on
Ximei Liang et al.
Ecotoxicology (London, England), 24(7-8), 1566-1573 (2015-04-22)
The presence of antibiotics including norfloxacin in the aquatic environment may cause adverse effects in non-target organisms. But the toxic mechanisms of fluoroquinolone to fish species are still not completely elucidated. Thus, it is essential to investigate the response of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service