Skip to Content
Merck
All Photos(1)

Key Documents

176966

Sigma-Aldrich

4-Quinolinecarboxaldehyde

97%

Synonym(s):

Cinchoninaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 G
$108.00
10 G
$354.00

$108.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2.5 G
$108.00
10 G
$354.00

About This Item

Empirical Formula (Hill Notation):
C10H7NO
CAS Number:
Molecular Weight:
157.17
Beilstein:
113072
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$108.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

impurities

3% lepidine

mp

45-52 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1ccnc2ccccc12

InChI

1S/C10H7NO/c12-7-8-5-6-11-10-4-2-1-3-9(8)10/h1-7H

InChI key

MGCGJBXTNWUHQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Quinolinecarboxaldehyde(cinchoninaldehyde) was used in the synthesis of lepidylamines[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cinchoninaldehyde and Aliphatic Amines?Preparation of Some Lepidylamines.
Phillips AP.
Journal of the American Chemical Society, 69(4), 865-866 (1947)
Alaa Naama-Amar et al.
Plants (Basel, Switzerland), 9(1) (2020-01-16)
Candidatus Phytoplasma, the causative agent of yellows disease, inflicts substantial damage on several hundred plant species including perennials and annual plants. The endophytic bacterium Frateuria defendens reduces the symptoms of yellows disease in a number of agricultural crops. One possible
Chunqing Li et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 228, 117720-117720 (2019-11-14)
It's of vital importance to detect heavy metals in environment and living cells. In this work, four near-infrared regions boron dipyrromethene (BODIPY) probes (QBPH, PBPH, QBP and PBP) are constructed based on two BODIPY precursors (QB, PB) for sensing of
Tamara Rodríguez-Cabo et al.
Analytical and bioanalytical chemistry, 407(20), 6159-6170 (2015-06-05)
Time-of-flight accurate mass spectrometry (TOF-MS), following a previous chromatographic (gas or liquid chromatography) separation step, is applied to the identification and structural elucidation of quinoline-like alkaloids in honey. Both electron ionization (EI) MS and positive electrospray (ESI+) MS spectra afforded

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service