176125
Trichloroisocyanuric acid
Synonym(s):
1,3,5-Trichloro-2,4,6-triazinetrione, Symclosene, TCICA, Trichlorocyanuric acid
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
form
powder
mp
249-251 °C (lit.)
SMILES string
ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O
InChI
1S/C3Cl3N3O3/c4-7-1(10)8(5)3(12)9(6)2(7)11
InChI key
YRIZYWQGELRKNT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Trichloroisocyanuric acid is used in the synthesis of 2-halomethylene-3-oxoketoximes.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Ox. Sol. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 2
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Trichloroisocyanuric acid: A safe and efficient oxidan
Organic Process Research & Development, 6(4), 384-393 (2002)
The Journal of organic chemistry, 78(17), 8872-8879 (2013-08-16)
A protocol for the N-bromosuccinimide (NBS)- and trichloroisocyanuric acid (TCCA)-mediated synthesis of novel 2-halomethylene-3-oxoketoximes via one-pot halogenation/oxidation of isoxazoline N-oxide derivatives is described here. The keto functionality of 3-ketoximes was selectively reduced by lithiumaluminum hydride to synthesize an unprecedented type
Acta chimica Slovenica, 60(4), 826-832 (2013-12-24)
In our study, the transformation of two most widely used UV filters, benzophenone-3 (BP3) and benzophenone-4 (BP4), in chlorinated water with disinfection reagents sodium hypochlorite (NaClO) and trichloroisocyanuric acid (TCCA) was studied. Based on the HPLC/MS and UV-Vis analysis the
Re: Effect of silver oxide/trichloroisocyanuric acid antimicrobial urinary drainage system on catheter-associated bacteriuria.
The Journal of urology, 141(5), 1213-1213 (1989-05-01)
Journal of hazardous materials, 161(2-3), 933-940 (2008-06-03)
High surface area (887.3m(2)/g) silica nanoparticles were synthesized using aerogel route and thereafter, characterized by N(2)-Brunauer-Emmet-Teller (BET), SEM and TEM techniques. The data indicated the formation of nanoparticles of silica in the size range of 24-75 nm with mesoporous characteristics.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service