Skip to Content
Merck
All Photos(1)

Key Documents

174335

Sigma-Aldrich

2-Amino-5-chlorobenzonitrile

98%

Synonym(s):

5-Chloroanthranilonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$93.80

$93.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$93.80

About This Item

Linear Formula:
H2NC6H3(Cl)CN
CAS Number:
Molecular Weight:
152.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$93.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

bp

132-135 °C/0.5 mmHg (lit.)

mp

96-99 °C (lit.)

SMILES string

Nc1ccc(Cl)cc1C#N

InChI

1S/C7H5ClN2/c8-6-1-2-7(10)5(3-6)4-9/h1-3H,10H2

InChI key

QYRDWARBHMCOAG-UHFFFAOYSA-N

Related Categories

General description

The vibrational spectra of 2-amino-5-chlorobenzonitrile was studied using density functinal theory[1].

Application

2-Amino-5-chlorobenzonitrile was used to build up the 2,2-dioxide-1H-2,1,3-benzothiadiazine ring system[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Krishnakumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(2), 465-470 (2008-04-22)
The vibrational spectra of 2-amino-5-chloro benzonitrile (ACB) have been obtained by density functional theory (DFT) calculations. Normal coordinate analysis has been carried out to support the vibrational analysis. The results were compared with the experimental values. With the help of
J W Corbett et al.
Bioorganic & medicinal chemistry letters, 10(2), 193-195 (2000-02-15)
Benzothiadiazine non-nucleoside reverse transcriptase inhibitors (NNRTIs) of HIV have been synthesized via a novel process to afford active inhibitors, with the most potent compound exhibiting an IC90 = 180 nM in a whole cell assay. The 2,2-dioxide-1H-2,1,3-benzothiadiazine ring system was
Jiayin Hu et al.
ChemSusChem, 11(24), 4219-4225 (2018-11-16)
The efficient transformation of CO2 into value-added chemicals with green, abundant, and cheap catalysts is an interesting and challenging topic in both green and sustainable chemistry. In this study, a series of salt-lake brines were used for the first time

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service