Skip to Content
Merck
All Photos(1)

Key Documents

174319

Sigma-Aldrich

p-Toluenesulfonhydrazide

~99%

Synonym(s):

p-Toluenesulfonic acid hydrazide, p-Toluenesulfonyl hydrazide, Tosylhydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NHNH2
CAS Number:
Molecular Weight:
186.23
Beilstein:
610130
EC Number:
MDL number:

Assay

~99%

availability

available only in EU

mp

103-108 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)NN

InChI

1S/C7H10N2O2S/c1-6-2-4-7(5-3-6)12(10,11)9-8/h2-5,9H,8H2,1H3

InChI key

ICGLPKIVTVWCFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Self-react. D

Supplementary Hazards

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda J Ejim et al.
Journal of medicinal chemistry, 50(4), 755-764 (2007-02-16)
The biosynthesis of methionine is an attractive antibiotic target given its importance in protein and DNA metabolism and its absence in mammals. We have performed a high-throughput screen of the methionine biosynthesis enzyme cystathionine beta-lyase (CBL) against a library of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service