Skip to Content
Merck
All Photos(1)

Key Documents

17028

Sigma-Aldrich

(3-Formyl-1-indolyl)acetic acid

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9NO3
CAS Number:
Molecular Weight:
203.19
Beilstein:
5431204
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

form

solid

reaction suitability

reagent type: cross-linking reagent

mp

197-200 °C (lit.)
197-200 °C

storage temp.

2-8°C

SMILES string

OC(=O)Cn1cc(C=O)c2ccccc12

InChI

1S/C11H9NO3/c13-7-8-5-12(6-11(14)15)10-4-2-1-3-9(8)10/h1-5,7H,6H2,(H,14,15)

InChI key

ZUUGBTJTGRTIFK-UHFFFAOYSA-N

Other Notes

Handle to synthesize an indolecarboxaldehyde resin by coupling to an amino-resin. Amines and derivatives can be immobilized by reductive amination of this aldehyde resin. Mild cleavage with 50% TFA[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.G. Estep et al.
The Journal of Organic Chemistry, 63, 5300-5300 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service