Skip to Content
Merck
All Photos(1)

Key Documents

164569

Sigma-Aldrich

Tritylium hexafluorophosphate

Synonym(s):

Triphenylcarbenium hexafluorophosphate, NSC 176018, Triphenylcarbonium hexafluorophosphate, Triphenylmethyl hexafluorophosphate, Trityl hexafluorophosphate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$161.00

$161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$161.00

About This Item

Linear Formula:
(C6H5)3C(PF6)
CAS Number:
Molecular Weight:
388.29
Beilstein:
4344297
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$161.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

mp

~150 °C (dec.) (lit.)

functional group

phenyl

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.c1ccc(cc1)[C+](c2ccccc2)c3ccccc3

InChI

1S/C19H15.F6P/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-7(2,3,4,5)6/h1-15H;/q+1;-1

InChI key

IBTFOFOFRZKIJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant of reagent involved in:
  • Si-H and Si-Si bond activeation at Pt of neutral and cationic silyl Pt bisphosphine complexes
  • Investigations of interactions with chromium acetylacetonate nitride mononuclear complexes
  • Synthesis of "push-pull" tropylium-fused aminoporphyrazine
  • Preparation of cycloheptatrienyl and tropylium calix[4]arenes
  • Synthesis of palladacycles containing chiral rhenium half-sandwich complex for use as a catalyst
  • Double ring-opening polyaddition of spiro orthoesters

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service