Skip to Content
Merck
All Photos(7)

Key Documents

164267

Sigma-Aldrich

3-Amino-2-naphthol

greener alternative

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC10H6OH
CAS Number:
Molecular Weight:
159.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

greener alternative product score

old score: 15
new score: 9
Find out more about DOZN™ Scoring

greener alternative product characteristics

Atom Economy
Design for Energy Efficiency
Use of Renewable Feedstocks
Reduce Derivatives
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

229-230 °C (lit.)

greener alternative category

SMILES string

Nc1cc2ccccc2cc1O

InChI

1S/C10H9NO/c11-9-5-7-3-1-2-4-8(7)6-10(9)12/h1-6,12H,11H2

InChI key

ZHVPTERSBUMMHK-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Atom economy”, “Design for Energy Efficiency”, “Use of Renewable Feedstock” and “Reduce Derivatives”. Click here to view its DOZN scorecard.

Application

3-Amino-2-naphthol was used in the synthesis of 2-naphthyl-3-NHSO2CF3, 1-naphthyl-5-NHSO2CF3, benzoquinolinequinone and 3-chloro-2-napthol.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service