Skip to Content
Merck
All Photos(1)

Key Documents

164100

Sigma-Aldrich

Ethyl acetoacetate sodium salt

Synonym(s):

Ethyl sodioacetoacetate, Ethyl sodium acetoacetate, Sodium ethyl acetoacetate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$167.00

$167.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$167.00

About This Item

Linear Formula:
CH3C(ONa)=CHCO2C2H5
CAS Number:
Molecular Weight:
152.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$167.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

90-110% (perchloric acid titration)

form

powder, crystals or chunks

reaction suitability

core: sodium

impurities

≤10% Acetic acid sodium salt

mp

168 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Na+].CCOC(=O)\C=C(/C)[O-]

InChI

1S/C6H10O3.Na/c1-3-9-6(8)4-5(2)7;/h4,7H,3H2,1-2H3;/q;+1/p-1/b5-4+;

InChI key

RYLPINAPVLRJBZ-FXRZFVDSSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service