Skip to Content
Merck
All Photos(3)

Key Documents

163023

Sigma-Aldrich

1,1-Dichloroethylene

contains 200 ppm MEHQ as inhibitor, 99%

Synonym(s):

Vinylidene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CCl2
CAS Number:
Molecular Weight:
96.94
Beilstein:
1733365
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

3.46 (vs air)

vapor pressure

9.68 psi ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

968 °F

contains

200 ppm MEHQ as inhibitor

expl. lim.

15.5 %

bp

30-32 °C (lit.)

mp

−122 °C (lit.)

density

1.213 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(Cl)=C

InChI

1S/C2H2Cl2/c1-2(3)4/h1H2

InChI key

LGXVIGDEPROXKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,1-Dichloroethylene can be used as a monomer to synthesize polymercomposite latexes by emulsion polymerization.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 1 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.2 °F

Flash Point(C)

-19 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan Chen et al.
Frontiers in microbiology, 9, 2306-2306 (2018-10-17)
Bioelectrochemical systems (BESs) are regarded as a promising approach for the enhanced dechlorination of chlorinated aliphatic hydrocarbons (CAHs). However, the electron distribution and transfer considering dechlorination, methanogenesis, and other bioprocesses in these systems are little understood. This study investigated the
Young Kim et al.
Biotechnology and bioengineering, 80(5), 498-508 (2002-10-02)
Batch kinetic and inhibition studies were performed for the aerobic cometabolism of 1,1,1-trichloroethane (1,1,1-TCA), 1,1-dichloroethylene (1,1-DCE), and 1,1-dichloroethane (1,1-DCA) by a butane-grown mixed culture. These chlorinated aliphatic hydrocarbons (CAHs) are often found together as cocontaminants in groundwater. The maximum degradation
William Slikker et al.
Toxicology and applied pharmacology, 201(3), 226-294 (2004-12-08)
Experience with dose response and mechanisms of toxicity has shown that multiple mechanisms may exist for a single agent along the continuum of the full dose-response curve. It is highly likely that critical, limiting steps in any given mechanistic pathway
Masarin Ban et al.
Toxicology, 226(2-3), 161-171 (2006-08-02)
Occupational allergy and asthma is a challenging issue in the developing countries. Chemicals inhaled in the workplaces may act not only as allergens but also as immune response modifiers, contributing to asthma exacerbation. In this study, we tested the adjuvant
Erik J Martin et al.
The Journal of pharmacology and experimental therapeutics, 313(1), 95-103 (2005-01-01)
1,1-Dichloroethylene (DCE) causes pulmonary injury that is characterized by necrosis of bronchiolar Clara cells. Mitochondria have been identified as an early target in the toxic response. Because mitochondria have been implicated in both necrotic and apoptotic cell death, we have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service