Skip to Content
Merck
All Photos(1)

Key Documents

161918

Sigma-Aldrich

2,2-Diphenylglycine

98%

Synonym(s):

α-amino-α-phenyl-Benzeneacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$178.00

$178.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$178.00

About This Item

Linear Formula:
H2NC(C6H5)2CO2H
CAS Number:
Molecular Weight:
227.26
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

$178.00


Check Cart for Availability

Request a Bulk Order

Product Name

2,2-Diphenylglycine, 98%

Quality Level

Assay

98%

form

powder or crystals

reaction suitability

reaction type: solution phase peptide synthesis

color

white to off-white

mp

245-247 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NC(C(O)=O)(c1ccccc1)c2ccccc2

InChI

1S/C14H13NO2/c15-14(13(16)17,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,15H2,(H,16,17)

InChI key

YBONNYNNFBAKLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service