Skip to Content
Merck
All Photos(1)

Key Documents

161896

Sigma-Aldrich

3,4-Dibenzyloxyphenethylamine hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2O)2C6H3CH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
369.88
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

131-133 °C (lit.)

functional group

amine
phenyl

SMILES string

Cl.NCCc1ccc(OCc2ccccc2)c(OCc3ccccc3)c1

InChI

1S/C22H23NO2.ClH/c23-14-13-18-11-12-21(24-16-19-7-3-1-4-8-19)22(15-18)25-17-20-9-5-2-6-10-20;/h1-12,15H,13-14,16-17,23H2;1H

InChI key

KXIZXPRLNNDQKS-UHFFFAOYSA-N

Application

3,4-Dibenzyloxyphenethylamine hydrochloride was used in the synthesis of tetrahydroisoquinoline derivatives containing 4(hexylureido)benzenesulfonamide, human β3 adrenergic receptor agonist. It was used as reagent in synthesis of the prodrugs of dopamine.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E R Parmee et al.
Bioorganic & medicinal chemistry letters, 10(20), 2283-2286 (2000-10-31)
Tetrahydroisoquinoline derivatives containing a 4-(hexylureido)benzenesulfonamide were examined as human beta3 adrenergic receptor (AR) agonists. Notably, 4,4-biphenyl derivative 9 was a 6 nM full agonist of the beta3 AR. Naphthyloxy compound 18 (beta3 EC50 = 78 nM) did not activate the
Lauri Peura et al.
Pharmaceutical research, 30(10), 2523-2537 (2013-10-19)
Drug delivery to the brain is impeded by the blood-brain barrier (BBB). Here, we attempted to enhance the brain uptake of cationic dopamine by utilizing the large amino acid transporter 1 (LAT1) at the BBB by prodrug approach. Three amino

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service