Skip to Content
Merck
All Photos(1)

Key Documents

161152

Sigma-Aldrich

2-Nitro-1-naphthol

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$144.00

$144.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$144.00

About This Item

Linear Formula:
O2NC10H6OH
CAS Number:
Molecular Weight:
189.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$144.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

mp

123-125 °C (lit.)

functional group

nitro

SMILES string

Oc1c(ccc2ccccc12)[N+]([O-])=O

InChI

1S/C10H7NO3/c12-10-8-4-2-1-3-7(8)5-6-9(10)11(13)14/h1-6,12H

InChI key

MUCCHGOWMZTLHK-UHFFFAOYSA-N

Application

2-Nitro-1-naphthol was employed as substrate for use in an amperometric 3-electrode system for the determination of alkaline phosphatase, the enzyme label most commonly used in electrochemical immunoassays.[1] It was also used in the preparation of:
  • di-(2-nitro-1-naphthyl) hydrogen phosphate[2]
  • 1-(2′-methyl-3′-indenyl)-2-naphthylamine and -2-naphthol (axially chiral ligands)[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, resolution and rates of racemisation of 1-(2'-methyl-3'-indenyl)-2-naphthylamine and-2-naphthol.
Baker RW and Taylor JA.
Tetrahedron Letters, 41(22), 4471-4473 (2000)
Alkaline phosphatase as a label for immunoassay using amperometric detection with a variety of substrates and an optimal buffer system.
Kreuzer MP, et al.
Analytica Chimica Acta, 393(1), 95-102 (1999)
The biochemistry of aromatic amines. 8. Synthesis and detection of di-(2-amino-1-naphthyl) hydrogen phosphate, a metabolite of 2-naphthylamine in dogs.
E Boyland et al.
The Biochemical journal, 78(1), 175-179 (1961-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service