159557
2-Chloro-5-methylphenol
99%
Synonym(s):
6-Chloro-m-cresol
Select a Size
About This Item
Recommended Products
Quality Level
Assay
99%
bp
196 °C (lit.)
mp
46-48 °C (lit.)
density
1.215 g/mL at 25 °C (lit.)
functional group
chloro
SMILES string
Cc1ccc(Cl)c(O)c1
InChI
1S/C7H7ClO/c1-5-2-3-6(8)7(9)4-5/h2-4,9H,1H3
InChI key
SMFHPCZZAAMJJO-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
177.8 °F - closed cup
Flash Point(C)
81 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service