Skip to Content
Merck
All Photos(1)

Key Documents

158356

Sigma-Aldrich

2-Benzimidazolemethanol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$177.00

$177.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$177.00

About This Item

Empirical Formula (Hill Notation):
C8H8N2O
CAS Number:
Molecular Weight:
148.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$177.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

171-175 °C (lit.)

functional group

hydroxyl

SMILES string

OCc1nc2ccccc2[nH]1

InChI

1S/C8H8N2O/c11-5-8-9-6-3-1-2-4-7(6)10-8/h1-4,11H,5H2,(H,9,10)

InChI key

IAJLTMBBAVVMQO-UHFFFAOYSA-N

Application

2-Benzimidazolemethanol was used in the preparation of 2-(chloromethyl)-1-(methylsulfonyl)benzimidazole via methanesulfonylation.[1] It was also used in the synthesis of 1H-benzimidazol-2-ylmethyl diethyl phosphate.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The methanesulfonylation of 2-benzimidazolemethanol and alpha-(2-benzimidazolyl)benzyl alcohol.
A J Charlson
Carbohydrate research, 29(1), 89-98 (1973-07-01)
Synthesis, acid-base and complexing properties with Cu (II), Co (II) and Zn (II) in aqueous solution of a novel 1H-benzimidazol-2-ylmethyl diethyl phosphate ligand: Comparison with other 2-substituted benzimidazole ligands
Kufelnicki A, et al.
Polyhedron, 53, 20-25 (2013)
Fangling Wu et al.
Talanta, 224, 121402-121402 (2021-01-01)
In this work, a sensitive, rapid, and matrix effect-free method for online simultaneous detection of benzimidazoles in animal products by in-tube solid-phase microextraction coupled with mass spectrometry (in-tube SPME-MS) was investigated. Herein, according to the chemical structures properites of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service