Skip to Content
Merck
All Photos(2)

Key Documents

157295

Sigma-Aldrich

Diethyl trans-1,2-cyclopropanedicarboxylate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$136.00
25 G
$652.00

$136.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$136.00
25 G
$652.00

About This Item

Linear Formula:
C3H4(CO2C2H5)2
CAS Number:
Molecular Weight:
186.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$136.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

70-75 °C/1 mmHg (lit.)

density

1.061 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)[C@@H]1C[C@H]1C(=O)OCC

InChI

1S/C9H14O4/c1-3-12-8(10)6-5-7(6)9(11)13-4-2/h6-7H,3-5H2,1-2H3/t6-,7-/m1/s1

InChI key

SXLDHZFJMXLFJU-RNFRBKRXSA-N

General description

Diethyl trans-1,2-cyclopropanedicarboxylate was obtained from the reaction of bis(acetonitrile)bis(diethyl fumarate)cobalt(0) with dibromomethane.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclopropanation of dimethyl fumarate and maleate with gem-dihalides catalyzed by Co (0)-or Ni (0)-complex and zinc.
Kanai H, et al.
Bulletin of the Chemical Society of Japan, 56(6), 1592-1597 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service