Skip to Content
Merck
All Photos(2)

Key Documents

156868

Sigma-Aldrich

(+)-N,N′-Diallyltartramide

≥99%

Synonym(s):

N,N′-Diallyl L-tartardiamide, N,N′-Diallyltartramide, DATD

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$86.40

$86.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$86.40

About This Item

Linear Formula:
[CH(OH)CONHCH2CH=CH2]2
CAS Number:
Molecular Weight:
228.25
Beilstein:
1712934
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$86.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

optical activity

[α]20/D +108°, c = 2.4 in H2O

mp

186-188 °C (lit.)

functional group

amide
hydroxyl

SMILES string

O[C@H]([C@@H](O)C(=O)NCC=C)C(=O)NCC=C

InChI

1S/C10H16N2O4/c1-3-5-11-9(15)7(13)8(14)10(16)12-6-4-2/h3-4,7-8,13-14H,1-2,5-6H2,(H,11,15)(H,12,16)/t7-,8-/m1/s1

InChI key

ZRKLEAHGBNDKHM-HTQZYQBOSA-N

General description

(+)-N,N′-Diallyltartramide can be used as a cross-linking agent during the preparation of hydrogels.[1][2]

Application

(+)-N,N′-Diallyltartramide can be used as a crosslinking agent:
  • In the polymerization of soluble polyacrylamide gels for electrophoresis applications.[3]
  • In the preparation of hydrogel containing cellulose, which is used as a component of an actuator capable of controlled soil irrigation.[4]
  • In the synthesis of dendronized polymers, which are used to prepare hydrogels containing ciprofloxacin for controlled drug release.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W R Springer
Analytical biochemistry, 195(1), 172-176 (1991-05-15)
A method is described in which individual proteins labeled with weak beta-emitting radionuclides, separated by polyacrylamide gel electrophoresis, and stained with silver are released from the gel by the use of the periodate soluble cross-linking agent N,N'-dialyltartardiamide. The radioactivity can
Electrophoresis and electrofocusing on polyacrylamide gel in the study of native macromolecules.
A Chrambach
Molecular and cellular biochemistry, 29(1), 23-46 (1980-01-16)
A novel gel based on an ionic complex from a dendronized polymer and ciprofloxacin: Evaluation of its use for controlled topical drug release
Garcia MC, et al.
Materials Science and Engineering, C, 69(2), 236-246 (2016)
Post-synthesis modification of hydrogels. Total and partial rupture of crosslinks: Formation of aldehyde groups and re-crosslinking of cleaved hydrogels.
Wolfel A, et al.
Polymer (2017)
Ashish Khaparde et al.
Journal of pharmaceutical and biomedical analysis, 181, 113099-113099 (2020-01-25)
A generic multi-component approach was designed to perform simultaneous in situ polymerization and ligand immobilization to develop affinity porous polymer based chromatography resin in a facile mode. This strategy exploits the regioselective ring opening reaction between epoxy group of monomer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service