Skip to Content
Merck
All Photos(1)

Key Documents

156752

Sigma-Aldrich

2,3-Dichloroanisole

≥97%

Synonym(s):

1,2-Dichloro-3-methoxybenzene

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$112.00

$112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$112.00

About This Item

Linear Formula:
Cl2C6H3OCH3
CAS Number:
Molecular Weight:
177.03
Beilstein:
1909166
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$112.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97%

form

solid

mp

31-33 °C (lit.)

functional group

chloro

SMILES string

COc1cccc(Cl)c1Cl

InChI

1S/C7H6Cl2O/c1-10-6-4-2-3-5(8)7(6)9/h2-4H,1H3

InChI key

HFEASCCDHUVYKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The product has been used to develop an analytical method for the determination of biogenic halogenated anisoles in the picogram/m3 range in air.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Führer et al.
Analytical and bioanalytical chemistry, 354(3), 333-343 (1996-01-01)
Halogenated anisoles (methyl-phenyl ethers) appear to be ubiquitous organic trace compounds in the environment. An analytical method is presented for analyzing the altogether 134 congeners of chloro-, bromo- and mixed bromochloro-anisoles on an isomer-specific basis in air. High volume sampling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service