Skip to Content
Merck
All Photos(1)

Key Documents

156116

Sigma-Aldrich

Tetrahydrofurfuryl acetate

97%

Synonym(s):

Tetrahydro-2-furylmethyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$59.80

$59.80


0 left in stock

Request a Bulk Order

Select a Size

Change View
50 ML
$59.80

About This Item

Empirical Formula (Hill Notation):
C7H12O3
CAS Number:
Molecular Weight:
144.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$59.80


0 left in stock

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.437 (lit.)

bp

194-195 °C/753 mmHg (lit.)

density

1.061 g/mL at 25 °C (lit.)

functional group

ester
ether

SMILES string

CC(=O)OCC1CCCO1

InChI

1S/C7H12O3/c1-6(8)10-5-7-3-2-4-9-7/h7H,2-5H2,1H3

InChI key

AAQDYYFAFXGBFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The radical-chain racemisation of (R)-tetrahydrofurfuryl acetate has been studied[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radical-chain racemisation of tetrahydrofurfuryl acetate under conditions of polarity-reversal catalysis: possible implications for the radical-induced strand cleavage of DNA.
Chemical Communications (Cambridge, England), 10, 1145-1146 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service