Skip to Content
Merck
All Photos(1)

Key Documents

155799

Sigma-Aldrich

4-Fluorostyrene

99%, contains tert-butylcatechol as inhibitor

Synonym(s):

1-Ethenyl-4-fluorobenzene, 1-Vinyl-4-fluorobenzene, p -Fluorostyrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC6H4F
CAS Number:
Molecular Weight:
122.14
Beilstein:
1634203
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

contains

tert-butylcatechol as inhibitor

refractive index

n20/D 1.514 (lit.)

bp

67 °C/50 mmHg (lit.)

density

1.024 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1ccc(C=C)cc1

InChI

1S/C8H7F/c1-2-7-3-5-8(9)6-4-7/h2-6H,1H2

InChI key

JWVTWJNGILGLAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oriol Planas et al.
Science (New York, N.Y.), 367(6475), 313-317 (2020-01-18)
Bismuth catalysis has traditionally relied on the Lewis acidic properties of the element in a fixed oxidation state. In this paper, we report a series of bismuth complexes that can undergo oxidative addition, reductive elimination, and transmetallation in a manner
Christoph Sprung et al.
Journal of the American Chemical Society, 137(5), 1916-1928 (2015-01-16)
A detailed and systematic polarized confocal fluorescence microscopy investigation is presented on three batches of large coffin-shaped ZSM-5 crystals (i.e., parent, steamed at 500 °C, and steamed at 700 °C). In total, six laser lines of different wavelength in the
Christopher L Rock et al.
Dalton transactions (Cambridge, England : 2003), 48(2), 461-467 (2018-11-30)
The phosphine-substituted α-diimine Ni precursor, (Ph2PPrDI)Ni, has been found to catalyze alkene hydrosilylation in the presence of Ph2SiH2 with turnover frequencies of up to 124 h-1 at 25 °C (990 h-1 at 60 °C). Moreover, the selective hydrosilylation of allylic

Articles

Fluorocarbon polymers exhibit increased thermal stability, hydrophobicity, lipophobicity, and chemical resistance compared to hydrocarbon analogs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service