Skip to Content
Merck
All Photos(1)

Key Documents

154296

Sigma-Aldrich

5-Bromo-2-methoxybenzaldehyde

99%

Synonym(s):

5-Bromo-o-anisaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(OCH3)CHO
CAS Number:
Molecular Weight:
215.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

116-119 °C (lit.)

SMILES string

COc1ccc(Br)cc1C=O

InChI

1S/C8H7BrO2/c1-11-8-3-2-7(9)4-6(8)5-10/h2-5H,1H3

InChI key

IJIBRSFAXRFPPN-UHFFFAOYSA-N

General description

The spectral properties of 5-Bromo-2-methoxybenzaldehyde have been studied[1].

Application

5-Bromo-2-methoxybenzaldehyde was used in the preparation of trans-5-bromo-2-methoxybenzylidenecyanoacetic acid[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

22. Stereochemistry of arylidenecyanoacetic acids and arylarylideneacetonitriles.
Journal of the Chemical Society, 119-124 (1953)
V Balachandran et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 106, 262-274 (2013-02-19)
The (FT-IR and FT-Raman) spectral properties of 5-bromo-2-methoxybenzaldehyde (BMB) are studied using density functional theory (DFT) employing B3LYP/6-311++G (d) and B3LYP/6-311++G (d,p) levels of theory. There are four conformers, C1, C2, C3, and C4 for this molecule. The computational results

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service