Skip to Content
Merck
All Photos(1)

Key Documents

154172

Sigma-Aldrich

4-Fluorophenethyl alcohol

97%

Synonym(s):

2-(4-Fluorophenyl)ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$219.00

$219.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$219.00

About This Item

Linear Formula:
FC6H4CH2CH2OH
CAS Number:
Molecular Weight:
140.15
Beilstein:
2354310
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$219.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.507 (lit.)

density

1.121 g/mL at 25 °C (lit.)

functional group

fluoro
hydroxyl

SMILES string

OCCc1ccc(F)cc1

InChI

1S/C8H9FO/c9-8-3-1-7(2-4-8)5-6-10/h1-4,10H,5-6H2

InChI key

MWUVGXCUHWKQJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Fluorophenethyl alcohol was used to prepare 1-fluoro-4-(2-iodoethyl)benzene[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of (?)-[4aα, 4bβ, 10bβ, 12aβ]-9-halogeno-2-methyl-1, 2, 3, 4, 4a, 4b, 5, 6, 10b, 11, 12, 12a-dodecahydronaphtho [2, 1-?] isoquinolines.
Patrick GL.
Journal of the Chemical Society. Perkin Transactions 1, 10, 1273-1279 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service