Skip to Content
Merck
All Photos(1)

Key Documents

15386

Sigma-Aldrich

Boc-Asp(OBzl)-OH

≥99.0% (sum of enantiomers, HPLC)

Synonym(s):

N-(tert-Butoxycarbonyl)-L-aspartic acid 4-benzyl ester, Boc-L-aspartic acid 4-benzyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$62.10

$62.10


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$62.10

About This Item

Linear Formula:
C6H5CH2OCOCH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
323.34
Beilstein:
2064127
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

$62.10


0 left in stock

Request a Bulk Order

Quality Level

Assay

≥99.0% (sum of enantiomers, HPLC)

optical activity

[α]20/D −20.0±1°, c = 2% in DMF

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

98-102 °C (lit.)

application(s)

peptide synthesis

SMILES string

[H][C@@](CC(=O)OCc1ccccc1)(NC(=O)OC(C)(C)C)C(O)=O

InChI

1S/C16H21NO6/c1-16(2,3)23-15(21)17-12(14(19)20)9-13(18)22-10-11-7-5-4-6-8-11/h4-8,12H,9-10H2,1-3H3,(H,17,21)(H,19,20)/t12-/m0/s1

InChI key

SOHLZANWVLCPHK-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole Niklas et al.
Dalton transactions (Cambridge, England : 2003), (1)(1), 154-162 (2006-12-13)
The amino acid derivative Boc-Asp-OBzl (Boc=N-butyloxycarbonyl; Asp=aspartic acid; Bzl=benzyl) was functionalized by coupling its carboxylate side chain to dipicolylamine. This yielded the tridentate nitrogen donor ligand Boc-Asp(Dpa)-OBzl (-OBzl). The compound -OBzl contains three different carbonyl groups: a tertiary amide linkage
Vitamin K1 dependent carboxylase: beta-carboxylation of t-butyloxycarbonylaspartic acid alpha-benzyl ester.
S E Hamilton et al.
Biochemical and biophysical research communications, 107(1), 246-249 (1982-07-16)
J J McTigue et al.
The Journal of biological chemistry, 259(7), 4272-4278 (1984-04-10)
The rat liver microsomal vitamin K-dependent carboxylase has been shown to carboxylate the aspartyl residue in N alpha-t-butyloxycarbonylaspartic acid alpha-benzyl ester to a beta-carboxyaspartyl residue. The enzyme also carboxylates aspartyl residues in low molecular weight peptides to beta-carboxyaspartyl residues, but
Tomohisa Sawada et al.
Nature communications, 10(1), 5687-5687 (2019-12-14)
Cavity creation is a key to the origin of biological functions. Small cavities such as enzyme pockets are created simply through liner peptide folding. Nature can create much larger cavities by threading and entangling large peptide rings, as learned from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service