Skip to Content
Merck
All Photos(1)

Key Documents

15372

Sigma-Aldrich

(R)-(+)-1-Boc-2-tert-butyl-3-methyl-4-imidazolidinone

≥99.0% (T)

Synonym(s):

(R)-(+)-1-(tert-Butoxycarbonyl)-2-tert-butyl-3-methyl-4-imidazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24N2O3
CAS Number:
Molecular Weight:
256.34
Beilstein:
4907143
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99.0% (T)

optical activity

[α]20/D +14±1°, c = 1% in methylene chloride

mp

67-69 °C
68-70 °C (lit.)

storage temp.

2-8°C

SMILES string

CN1[C@H](N(CC1=O)C(=O)OC(C)(C)C)C(C)(C)C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Chiral glycine derivative used for the synthesis of amino acids

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Gander-Coquoz et al.
Helvetica Chimica Acta, 71, 224-224 (1988)
D. Seebach et al.
Helvetica Chimica Acta, 70, 237-237 (1987)
R. Fitzi et al.
Tetrahedron, 44, 5277-5277 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service