Skip to Content
Merck
All Photos(2)

Key Documents

153230

Sigma-Aldrich

N-(2-Amino-4-chlorophenyl)anthranilic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$158.00

$158.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$158.00

About This Item

Linear Formula:
H2NC6H3(Cl)NHC6H4CO2H
CAS Number:
Molecular Weight:
262.69
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$158.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

191 °C (dec.) (lit.)

functional group

carboxylic acid
chloro

SMILES string

Nc1cc(Cl)ccc1Nc2ccccc2C(O)=O

InChI

1S/C13H11ClN2O2/c14-8-5-6-12(10(15)7-8)16-11-4-2-1-3-9(11)13(17)18/h1-7,16H,15H2,(H,17,18)

InChI key

HMVPMZFEYCGSTC-UHFFFAOYSA-N

Related Categories

Application

N-(2-Amino-4-chlorophenyl)anthranilic acid may be used in chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young Jin Seong et al.
Antimicrobial agents and chemotherapy, 64(7) (2020-05-13)
The rise of extensively drug-resistant and multidrug-resistant strains of Neisseria gonorrhoeae has occurred in parallel with the increasing demand for new drugs. However, the current methods of drug discovery are burdened with rigorous assessments and require more time than can

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service