Skip to Content
Merck
All Photos(3)

Key Documents

152811

Sigma-Aldrich

3,4-Dichlorophenylhydrazine hydrochloride

technical grade, 90%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$356.00

$356.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$356.00

About This Item

Linear Formula:
Cl2C6H3NHNH2 · HCl
CAS Number:
Molecular Weight:
213.49
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$356.00


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

Assay

90%

mp

230 °C (dec.) (lit.)

functional group

chloro
hydrazine

SMILES string

Cl.NNc1ccc(Cl)c(Cl)c1

InChI

1S/C6H6Cl2N2.ClH/c7-5-2-1-4(10-9)3-6(5)8;/h1-3,10H,9H2;1H

InChI key

WIKZRYSGQPZTMM-UHFFFAOYSA-N

Application

3,4-Dichlorophenylhydrazine hydrochloride was used to prepare ethyl 2-(3,4-dichlorophenyl)hydrazinecarboxylate[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Hirose et al.
Angewandte Chemie (International ed. in English), 52(17), 4613-4617 (2013-03-08)
Aerobic recycling: A catalytic amount of a hydrazine reagent is sufficient to promote Mitsunobu reactions in the presence of triphenylphosphine, an iron catalyst, and air. The active form of the catalyst, an azo species, can be readily generated by iron-catalyzed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service