Skip to Content
Merck
All Photos(2)

Key Documents

149160

Sigma-Aldrich

3-Methylanisole

99%

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$117.00

$117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$117.00

About This Item

Linear Formula:
CH3C6H4OCH3
CAS Number:
Molecular Weight:
122.16
Beilstein:
907023
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$117.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.513 (lit.)

bp

175-176 °C (lit.)

density

0.969 g/mL at 25 °C (lit.)

SMILES string

COc1cccc(C)c1

InChI

1S/C8H10O/c1-7-4-3-5-8(6-7)9-2/h3-6H,1-2H3

InChI key

OSIGJGFTADMDOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methylanisole was used to study the functionalized core of calyciphylline A-type alkaloids[1].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Filippo Sladojevich et al.
Organic letters, 13(19), 5132-5135 (2011-09-09)
An efficient, robust, and scalable strategy to access the functionalized core of calyciphylline A-type alkaloids has been developed starting from commercially available 3-methylanisole. Key features of this approach are an intramolecular Michael addition/allylation sequence and a ring-closing metathesis step.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service