Skip to Content
Merck
All Photos(1)

Documents

147451

Sigma-Aldrich

Tetraphenylarsonium(V) chloride hydrochloride hydrate

Synonym(s):

Tetraphenylarsonium chloride hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)4AsCl·HCl·xH2O
CAS Number:
Molecular Weight:
455.25 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: arsenic
reagent type: catalyst

mp

211-213 °C (lit.)

SMILES string

O.Cl.[Cl-].c1ccc(cc1)[As+](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C24H20As.2ClH.H2O/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24;;;/h1-20H;2*1H;1H2/q+1;;;/p-1

InChI key

HRCBNZHNCWEFFR-UHFFFAOYSA-M

General description

Tetraphenylarsonium(V) chloride hydrochloride hydrate is commonly used as a reagent in titration.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potentiometric titration of perchlorate with tetraphenylarsonium chloride and a perchlorate ion specific electrode
Baczuk RJ, et al.
Analytical Chemistry, 40(4), 685-689 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service