Skip to Content
Merck
All Photos(3)

Key Documents

146757

Sigma-Aldrich

2,4-Dichlorobenzaldehyde

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$48.10
100 G
$91.30

$48.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$48.10
100 G
$91.30

About This Item

Linear Formula:
Cl2C6H3CHO
CAS Number:
Molecular Weight:
175.01
Beilstein:
471601
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$48.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

bp

233 °C (lit.)

mp

64-69 °C (lit.)

functional group

aldehyde
chloro

SMILES string

[H]C(=O)c1ccc(Cl)cc1Cl

InChI

1S/C7H4Cl2O/c8-6-2-1-5(4-10)7(9)3-6/h1-4H

InChI key

YSFBEAASFUWWHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dichlorobenzaldehyde on condensation with 4-amino-5-mercapto-3-methyl/ethyl-1,2,4-triazole yields bidentate Schiff base ligands and their metal complexes are potential antimicrobial agents[1].

Application

2,4-Dichlorobenzaldehyde was used in the solid-phase synthesis of a series of indole-based peptoids[2].

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

275.0 °F - closed cup

Flash Point(C)

135 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiran Singh et al.
Bioinorganic chemistry and applications, 2011, 901716-901716 (2012-01-05)
A series of cobalt, nickel, copper, and zinc complexes of bidentate Schiff bases derived from the condensation reaction of 4-amino-5-mercapto-3-methyl/ethyl-1,2,4-triazole with 2,4-dichlorobenzaldehyde were synthesized and tested as antimicrobial agents. The synthesized Schiff bases and their metal complexes were characterized with
Martina Quintanar-Audelo et al.
Journal of medicinal chemistry, 50(24), 6133-6143 (2007-11-08)
The vanilloid receptor subunit 1, or transient receptor potential vanilloid 1 (TRPV1), integrates physical and chemical stimuli in the peripheral nervous system, playing a key role in inflammatory pain. Identification of potent TRPV1 antagonists is thus an important goal of
Adnan Ashraf et al.
European journal of medicinal chemistry, 159, 282-291 (2018-10-09)
Chalcones and 1,2-benzothiazines are two important classes of bioactive compounds, each scaffold endowed with diverse pharmacological activities. Combining both of these pharmacophores in a single molecule was aimed to yield multi-modal agents. Herein, we report a series of hybrid compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service