Skip to Content
Merck
All Photos(3)

Key Documents

145963

Sigma-Aldrich

Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$33.70
25 G
$133.00
100 G
$501.00

$33.70


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$33.70
25 G
$133.00
100 G
$501.00

About This Item

Empirical Formula (Hill Notation):
C8H9ClN2O2S
CAS Number:
Molecular Weight:
232.69
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$33.70


0 left in stock

Request a Bulk Order

Assay

98%

form

powder

mp

60-63 °C (lit.)

functional group

chloro
ester
thioether

storage temp.

2-8°C

SMILES string

CCOC(=O)c1cnc(SC)nc1Cl

InChI

1S/C8H9ClN2O2S/c1-3-13-7(12)5-4-10-8(14-2)11-6(5)9/h4H,3H2,1-2H3

InChI key

SNNHLSHDDGJVDM-UHFFFAOYSA-N

Application

Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate was used in the synthesis of pyrimidinopyridones, potential inhibitors of the FMS tyrosine kinase[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Huang et al.
Bioorganic & medicinal chemistry letters, 18(7), 2355-2361 (2008-03-18)
A series of pyrimidinopyridones has been designed, synthesized and shown to be potent and selective inhibitors of the FMS tyrosine kinase. Introduction of an amide substituent at the 6-position of the pyridone core resulted in a significant potency increase. Compound

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service