Skip to Content
Merck
All Photos(1)

Key Documents

145831

Sigma-Aldrich

Allylamine

98%

Synonym(s):

2-Propen-1-ylamine, 3-Amino-1-propene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2NH2
CAS Number:
Molecular Weight:
57.09
Beilstein:
635703
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
98%

vapor density

2 (vs air)

vapor pressure

4.09 psi ( 20 °C)

Assay

98%

form

liquid

expl. lim.

22 %

refractive index

n20/D 1.420 (lit.)

bp

53 °C (lit.)

mp

−88 °C (lit.)

solubility

alcohol: miscible
chloroform: miscible
diethyl ether: miscible
water: miscible

density

0.761 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NCC=C

InChI

1S/C3H7N/c1-2-3-4/h2H,1,3-4H2

InChI key

VVJKKWFAADXIJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Allylamine can be used:
  • To prepare a chemical gradient through plasma co-polymerization of octadiene and allylamine polymer on nanoporous alumina.
  • As a monomer to prepare plasma polymerized allylamine films in the study of their reactivity on Au and Si.
  • In a direct monocarboxymethylation reaction to yield N-formyl glycine derivatives.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemical reactivity of plasma polymerized allylamine (PPAA) thin films on Au and Si: Study of the thickness influence and aging of the films.
Moreau N, et al.
Surface and Coatings Technology, 205, S462-S465 (2011)
Versatile gradients of chemistry, bound ligands and nanoparticles on alumina nanopore arrays
Michelmore A, et al.
Nanotechnology, 22(41), 415601-415601 (2011)
A mild and efficient method for the one-pot monocarboxymethylation of primary amines
Gibbs Timothy JK, et al.
Synlett, 2007(10), 1573-1576 (2007)
Chemical reactivity of plasma polymerized allylamine (PPAA) thin films on Au and Si: Study of the thickness influence and aging of the films
Moreau N, et al.
Surface and Coatings Technology, 205(41), S462-S465 (2011)
Synlett, 1573-1573 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service