Skip to Content
Merck
All Photos(1)

Documents

145394

Sigma-Aldrich

4-Phenylbutylamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)4NH2
CAS Number:
Molecular Weight:
149.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.519 (lit.)

bp

123-124 °C/17 mmHg (lit.)

density

0.944 g/mL at 25 °C (lit.)

SMILES string

NCCCCc1ccccc1

InChI

1S/C10H15N/c11-9-5-4-8-10-6-2-1-3-7-10/h1-3,6-7H,4-5,8-9,11H2

InChI key

AGNFWIZBEATIAK-UHFFFAOYSA-N

Gene Information

human ... AOC3(8639)
mouse ... Aoc3(11754)

Application

4-Phenylbutylamine was used in direct solvent-free amination of multi-walled carbon nanotubes. It was employed as internal standard during solid-phase micro-extraction by GC technique for detection of amphetamines in urine.

Biochem/physiol Actions

4-Phenylbutylamine is a competitive inhibitor of recombinant human liver monoamine oxidase A.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R K Nandigama et al.
Biochemistry, 39(49), 15258-15265 (2000-12-07)
The interaction of recombinant human liver monoamine oxidase A (MAO A) with a series of phenethylamine substrate analogues has been investigated by steady-state and stopped-flow kinetic techniques. Substrate analogues with para substituents exhibit large deuterium kinetic isotope effect on k(cat)
Ting Wang et al.
Journal of analytical toxicology, 44(6), 596-600 (2019-11-30)
In this study, the relationships between the concentrations of R/S-methamphetamine (MA) and its metabolite R/S-amphetamine (AP), the AP/MA ratio in hair samples, and MA dependence were investigated by performing segmental hair analysis in MA users. Authentic hair samples collected from
Lizhu Chen et al.
Drug testing and analysis, 11(6), 898-905 (2019-01-08)
Chiral analysis is a crucial way to differentiate selegiline (SG) intake from drug abuse. Oral fluid (OF) has been successfully used as an alternative matrix for blood testing in several pharmacokinetic studies. The aim of this study is to describe
Elena V Basiuk et al.
Journal of nanoscience and nanotechnology, 5(6), 984-990 (2005-08-03)
We performed direct solvent-free amination of multi-walled carbon nanotubes (MWCNTs) with nonylamine, dodecylamine, octadecylamine, 4-phenylbutylamine and 1,8-ocanediamine at a temperature of 150-170 degrees C and reduced pressure. Thermogravimetric analysis and temperature-programmed desorption-mass spectrometry revealed that a major amine fraction decomposes
Sho Kataoka et al.
Langmuir : the ACS journal of surfaces and colloids, 34(14), 4166-4172 (2018-03-20)
Hybrid organic-inorganic layered perovskites are typically nonporous solids. However, the incorporation of silsesquioxanes with a cubic cage structure as interlayer materials creates micropores between the perovskite layers. In this study, we increase in the micropore volume in layered perovskites by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service