Skip to Content
Merck
All Photos(1)

Key Documents

145246

Sigma-Aldrich

2H-Pyrido[3,2-b]-1,4-oxazin-3(4H)-one

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$173.00

$173.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$173.00

About This Item

Empirical Formula (Hill Notation):
C7H6N2O2
CAS Number:
Molecular Weight:
150.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$173.00


0 left in stock

Request a Bulk Order

Assay

95%

form

liquid

mp

204-206 °C (lit.)

SMILES string

O=C1COc2cccnc2N1

InChI

1S/C7H6N2O2/c10-6-4-11-5-2-1-3-8-7(5)9-6/h1-3H,4H2,(H,8,9,10)

InChI key

ANHQLUBMNSSPBV-UHFFFAOYSA-N

Application

2H-Pyrido[3,2-b]-1,4-oxazin-3(4H)-one was used in the synthesis of quinazolinbenzoxazine derivatives[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margarete von Wantoch Rekowski et al.
Bioorganic & medicinal chemistry, 18(3), 1288-1296 (2009-12-29)
Soluble guanylyl cyclase (sGC) is an ubiquitously expressed enzyme that generates the second messenger cGMP and hence, leads to a number of physiological responses including vasodilation, inhibition of platelet aggregation and neurotransmission. Whilst many activating and stimulating modulators of sGC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service