Skip to Content
Merck
All Photos(2)

Key Documents

144452

Sigma-Aldrich

tert-Butyl isocyanate

97%

Synonym(s):

2-isocyanato-2-methylpropane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$51.50
25 G
$152.00
100 G
$508.00

$51.50


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$51.50
25 G
$152.00
100 G
$508.00

About This Item

Linear Formula:
(CH3)3CN=C=O
CAS Number:
Molecular Weight:
99.13
Beilstein:
969479
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$51.50


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.386 (lit.)

bp

85-86 °C (lit.)

density

0.868 g/mL at 25 °C (lit.)

functional group

amine
isocyanate

SMILES string

CC(C)(C)N=C=O

InChI

1S/C5H9NO/c1-5(2,3)6-4-7/h1-3H3

InChI key

MGOLNIXAPIAKFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl isocyanate undergoes [2+2] cycloaddition reaction with phosphagermaallene to yield 1-oxa-2-germacyclobutane[1]. It also undergoes cycladdition reaction with molybdenum dioxo bis(aryloxide) complex to form novel 16-electron molybdenum oxo-imido bis(aryloxide) complex[2]. It reacts with aluminum dihydride in the presence of trace amounts of water to yield terminal hydroxide containing dinuclear alumoxane[3].

Application

tert-Butyl isocyanate can be used:
  • In the preparation of isocyanate-treated graphite oxides (iGOs) which can be exfoliated in polar aprotic solvents to obtain graphene oxide nanoplatelets.[4]
  • To modify metal-organic frameworks, postsynthetically, to obtain microporous urea-functionalized frameworks.[5]
  • To the synthesis of terminal hydroxide containing dinuclear alumoxane from aluminum dihydride.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L W Wattenberg
Cancer research, 41(8), 2991-2994 (1981-08-01)
The effects of sodium cyanate, tert-butyl isocyanate, and benzyl isothiocyanate on carcinogen-induced neoplasia were studied in experiments in which the test compound was fed starting 1 week following completion of carcinogen administrations. Under these conditions, all three test compounds exerted
Ying Peng et al.
Inorganic chemistry, 43(4), 1217-1219 (2004-02-18)
A novel terminal hydroxide containing dinuclear alumoxane LAl(OH)OAlL(OCH=N-tBu) (3; L = HC(CMeNAr)2, Ar = 2,6-iPr2C6H3) was prepared by treatment of aluminum dihydride LAlH2 (1) and tert-butyl isocyanate in the presence of trace amounts of water and alternatively from 1 and
Dumitru Ghereg et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(45), 12763-12772 (2011-10-01)
Phosphagermaallene Tip(tBu)Ge=C=PMes* 1 (Tip=2,4,6-triisopropylphenyl, Mes*=2,4,6-tri-tert-butylphenyl) reacts with phenyl isocyanate and tert-butyl isocyanate by a [2+2] cycloaddition that involves the Ge=C and C=O double bonds to afford 1-oxa-2-germacyclobutanes 2 and 3. With N,N'-dicyclohexylcarbodiimide, a [2+2] cycloaddition is observed between the Ge=C
Warren B Cross et al.
Inorganic chemistry, 45(11), 4556-4561 (2006-05-23)
The novel 16-electron molybdenum oxo-imido bis(aryloxide) complexes [Mo(NtBu)(O)(2,6-Me2C6H3O)2(py)] (1) and [Mo(NtBu)(O)(2,6-iPr2C6H3O)2(py)] (2) have been prepared by the salt elimination reactions of [Mo(NtBu)(O)Cl2(DME)] with the appropriate lithium aryloxide and from the cycloaddition reactions of tert-butyl isocyanate with the appropriate molybdenum dioxo
Emily Dugan et al.
Chemical communications (Cambridge, England), (29)(29), 3366-3368 (2008-07-18)
Isoreticular metal-organic framework-3 (IRMOF-3) has been postsynthetically modified with isocyanates to generate unprecedented, microporous urea-functionalized frameworks.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service