Skip to Content
Merck
All Photos(2)

Documents

144169

Sigma-Aldrich

2,3-Hexanedione

technical grade, 90%

Synonym(s):

Acetyl butyryl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COCOCH3
CAS Number:
Molecular Weight:
114.14
Beilstein:
1699896
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

vapor density

3.9 (vs air)

vapor pressure

10 mmHg ( 20 °C)

Assay

90%

form

liquid

refractive index

n20/D 1.412 (lit.)

bp

128 °C (lit.)

density

0.934 g/mL at 25 °C (lit.)

SMILES string

CCCC(=O)C(C)=O

InChI

1S/C6H10O2/c1-3-4-6(8)5(2)7/h3-4H2,1-2H3

InChI key

MWVFCEVNXHTDNF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2,3-Hexanedione is a major component of the male-produced aggregation pheromone of Anelaphus inflaticollis Chemsak. It reacts with ethylenediamine to yield macrocyclic tetradentate 12-membered nitrogen donor (N4) ligand.

Application

2,3-Hexanedione was used to evaluate the influence of xanthan concentration on the release of aroma compounds in xanthan-thickened food model systems.

Pictograms

FlameHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT RE 2 Inhalation

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulekh Chandra et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(13), 3079-3085 (2004-10-13)
Complexes of Cr(III), Co(II), Ni(II) and Cu(II) containing a novel macrocyclic tetradentate nitrogen donor (N4) ligand prepared via reaction of 2,3-hexanedione and ethylenediamine has been prepared and characterized. The newly synthesized ligand (L) and its complexes have been characterized on
A M Ray et al.
Environmental entomology, 38(5), 1462-1466 (2009-10-15)
We report the identification and field bioassays of a major component of the male-produced aggregation pheromone of Anelaphus inflaticollis Chemsak, an uncommon desert cerambycine beetle. Male A. inflaticollis produced a sex-specific blend of components that included (R)-3-hydroxyhexan-2-one, (S)-2-hydroxyhexan-3-one, 2,3-hexanedione, and
Egle Bylaite et al.
Journal of agricultural and food chemistry, 53(9), 3577-3583 (2005-04-28)
The influence of xanthan concentration (0, 0.02, 0.1, 0.4, and 0.8% w/w) and bulk viscosity on the release of 20 aroma compounds of different chemical classes (5 aldehydes, 4 esters, 5 ketones, 3 alcohols, and 3 terpenes) was evaluated in
Emerson S Lacey et al.
Journal of chemical ecology, 30(8), 1493-1507 (2004-11-13)
This is the first fully verified report of an aggregation pheromone produced by a cerambycid beetle species. Field bioassays with adult Neoclytus acuminatus acuminatus (F.) (Coleoptera: Cerambycidae) revealed that males produce a pheromone that attracts both sexes. Extracts of odors
Molecular mechanism of hexane neuropathy: significant differences in pharmacokinetics between 2.3-, 2.4-, and 2.5-hexanedione.
K Iwasaki et al.
Industrial health, 22(3), 177-187 (1984-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service