Skip to Content
Merck
All Photos(1)

Documents

143901

Sigma-Aldrich

Diethyl dimethylmalonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(CO2C2H5)2
CAS Number:
Molecular Weight:
188.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.412 (lit.)

bp

192 °C (lit.)

density

0.991 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(C)(C)C(=O)OCC

InChI

1S/C9H16O4/c1-5-12-7(10)9(3,4)8(11)13-6-2/h5-6H2,1-4H3

InChI key

UELKSYXXNGHTSE-UHFFFAOYSA-N

General description

Diethyl dimethylmalonate forms adducts with α,α,α′,α′-tetraaryl-1,3-dioxolane-4,5-dimethanol based titanium catalysts.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TADDOL-Based Titanium Catalysts and Their Adducts: Understanding Asymmetric Catalysis of Diels-Alder Reactions.
Haase C, et al.
The Journal of Organic Chemistry, 60(6), 1777-1787 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service