Assay
97%
mp
119-120 °C (dec.) (lit.)
SMILES string
CCCCN(N=O)C(=N)N[N+]([O-])=O
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 1B - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of bacteriology, 156(1), 6-12 (1983-10-01)
The mutagenic and toxic effects of a series of N-alkyl-N'-nitro-N-nitrosoguanidines were examined in Escherichia coli K-12. The role of nucleotide excision repair, the SOS response, and the adaptive response in both the reduction and the production of the biological effects
Journal of bacteriology, 177(6), 1511-1519 (1995-03-01)
Escherichia coli has two O6-methylguanine DNA methyltransferases that repair alkylation damage in DNA and are encoded by the ada and ogt genes. The ada gene of E. coli also regulates the adaptive response to alkylation damage. The closely related species
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service