Skip to Content
Merck
All Photos(1)

Key Documents

13955

Sigma-Aldrich

Benzyltriethylammonium iodide

≥98.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(I)(C2H5)3
CAS Number:
Molecular Weight:
319.22
Beilstein:
3918218
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

≥98.0% (AT)

SMILES string

[I-].CC[N+](CC)(CC)Cc1ccccc1

InChI

1S/C13H22N.HI/c1-4-14(5-2,6-3)12-13-10-8-7-9-11-13;/h7-11H,4-6,12H2,1-3H3;1H/q+1;/p-1

InChI key

JWLJBISFJGEYMT-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luís Almeida et al.
Immunity, 54(1), 68-83 (2020-11-26)
While antibiotics are intended to specifically target bacteria, most are known to affect host cell physiology. In addition, some antibiotic classes are reported as immunosuppressive for reasons that remain unclear. Here, we show that Linezolid, a ribosomal-targeting antibiotic (RAbo), effectively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service