Skip to Content
Merck
All Photos(3)

Key Documents

138150

Sigma-Aldrich

2,3-Dimethylbenzoic acid

98%

Synonym(s):

vic.-o-Xylylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$54.50

$54.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$54.50

About This Item

Linear Formula:
(CH3)2C6H3CO2H
CAS Number:
Molecular Weight:
150.17
Beilstein:
971590
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$54.50


Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

144-146 °C (lit.)

functional group

carboxylic acid

SMILES string

Cc1cccc(C(O)=O)c1C

InChI

1S/C9H10O2/c1-6-4-3-5-8(7(6)2)9(10)11/h3-5H,1-2H3,(H,10,11)

InChI key

RIZUCYSQUWMQLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2,3-Dimethylbenzoic acid shows high binding affinity with antenna-specific odorant-binding protein of the alfalfa plant bug, Adelphocoris lineolatus[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W A Duetz et al.
Journal of bacteriology, 176(8), 2354-2361 (1994-04-01)
The TOL catabolic genes in Pseudomonas putida (pWW0) are clustered in the upper operon, encoding enzymes for the conversion of toluene and xylenes to benzoate and toluates, and the meta-cleavage operon, encoding enzymes for the conversion of the benzoate and
Metabolic fate of 3,4,5- and 2,3,5-trimethylphenyl methylcarbamates, the major constituents in Landrin insecticide.
M Slade et al.
Journal of agricultural and food chemistry, 18(3), 467-474 (1970-05-01)
S-Y Wang et al.
Insect molecular biology, 22(2), 172-182 (2013-01-09)
Olfaction plays an important role in insects' survival and reproduction. Odorant-binding proteins (OBPs) are considered to be one of the crucial proteins in the insect olfactory pathway. In this study, an antenna-specific OBP of the alfalfa plant bug, Adelphocoris lineolatus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service