Skip to Content
Merck
All Photos(1)

Key Documents

136727

Sigma-Aldrich

3-Chlorobenzyl bromide

97%

Synonym(s):

α-Bromo-3-chlorotoluene

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$59.60
50 G
$178.00

$59.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$59.60
50 G
$178.00

About This Item

Linear Formula:
ClC6H4CH2Br
CAS Number:
Molecular Weight:
205.48
Beilstein:
636504
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$59.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.588 (lit.)

bp

109-110 °C/12 mmHg (lit.)

density

1.565 g/mL at 25 °C (lit.)

functional group

bromo
chloro

SMILES string

Clc1cccc(CBr)c1

InChI

1S/C7H6BrCl/c8-5-6-2-1-3-7(9)4-6/h1-4H,5H2

InChI key

LZIYAIRGDHSVED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

3-Chlorobenzyl bromide reacts with aminoethanol and NaH to yield aminoethyl 3-chlorobenzyl ether[1].

Application

3-Chlorobenzyl bromide was used in the synthesis of symmetrical and unsymmetrical benzyl thioethers[2]. It was used as starting reagent during the synthesis of 1-(3-chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)-1H-benzimidazole dihydrochloride[3].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective inactivation of monoamine oxidase B by aminoethyl 3-chlorobenzyl ether.
Ding CZ and Silverman RB.
Bioorganic & Medicinal Chemistry, 3(10), 2077-2078 (1993)
Convenient and robust one-pot synthesis of symmetrical and unsymmetrical benzyl thioethers from benzyl halides using thiourea.
Eccles KS, et al.
ARKIVOC (Gainesville, FL, United States), ix, 216-218 (2010)
Jason Z Vlahakis et al.
Bioorganic & medicinal chemistry, 21(21), 6788-6795 (2013-09-12)
Several analogs based on the lead structure of 1-(4-chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)-1H-benzimidazole (clemizole) were synthesized and evaluated as novel inhibitors of heme oxygenase (HO). Many of the compounds were found to be potent and highly selective for the HO-2 isozyme (constitutive), and had

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service