Skip to Content
Merck
All Photos(1)

Key Documents

135984

Sigma-Aldrich

Methyl 5-chlorovalerate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$235.00

$235.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$235.00

About This Item

Linear Formula:
Cl(CH2)4CO2CH3
CAS Number:
Molecular Weight:
150.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$235.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

106-107 °C/38 mmHg (lit.)

density

1.047 g/mL at 25 °C (lit.)

functional group

chloro
ester

SMILES string

COC(=O)CCCCCl

InChI

1S/C6H11ClO2/c1-9-6(8)4-2-3-5-7/h2-5H2,1H3

InChI key

JAVHFVJOWIQHII-UHFFFAOYSA-N

General description

Methyl 5-chlorovalerate undergoes gas phase elimination over the temperature range of 419.6–472.1°C and pressure range of 45–108 torr via intimate ion pair type mechanism[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The intimate ion pair mechanism in the maximally inhibited elimination kinetics of methyl 4-chlorobutyrate and methyl 5-chlorovalerate in the gas phase.
Chuchani G, et al.
International Journal of Chemical Kinetics, 18(2), 203-213 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service