Skip to Content
Merck
All Photos(1)

Key Documents

135119

Sigma-Aldrich

Triethyl phosphonoformate

98%

Synonym(s):

Ethyl diethoxyphospinylformate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(O)COOC2H5
CAS Number:
Molecular Weight:
210.16
Beilstein:
1781949
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.423 (lit.)

bp

135 °C/12 mmHg (lit.)

density

1.11 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)P(=O)(OCC)OCC

InChI

1S/C7H15O5P/c1-4-10-7(8)13(9,11-5-2)12-6-3/h4-6H2,1-3H3

InChI key

NOJFJZZMRDSOLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and antiviral properties of sterol phosphonoformates.
J M Bailey et al.
Biochemical Society transactions, 22(3), 353S-353S (1994-08-01)
Phosphonoformate inhibits synthesis of Epstein-Barr virus (EBV) capsid antigen and transformation of human cord blood lymphocytes by EBV.
M Margalith et al.
Virology, 102(1), 226-230 (1980-04-15)
M Margalith et al.
Molecular and cellular biochemistry, 43(2), 97-103 (1982-03-19)
Three potential inhibitors of reverse transcriptase activities, phosphonoformate (PF), phosphonoacetate (PAA), and ethyl-diethyl phosphonoformate (Et-PF), were compared in this study. Only PF was found to inhibit the DNA polymerase activity of the purified reverse transcriptase of Moloney murine leukemia virus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service