Skip to Content
Merck
All Photos(1)

Key Documents

134309

Sigma-Aldrich

trans-2-Phenyl-1-cyclopropanecarbonyl chloride

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C3H4COCl
CAS Number:
Molecular Weight:
180.63
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.556 (lit.)

bp

108-110 °C/2 mmHg (lit.)

density

1.163 g/mL at 25 °C (lit.)

functional group

acyl chloride
phenyl

SMILES string

ClC(=O)[C@@H]1C[C@H]1c2ccccc2

InChI

1S/C10H9ClO/c11-10(12)9-6-8(9)7-4-2-1-3-5-7/h1-5,8-9H,6H2/t8-,9+/m0/s1

InChI key

SODZBMGDYKEZJG-DTWKUNHWSA-N

Application

trans-2-Phenyl-1-cyclopropanecarbonyl chloride was used in the synthesis of series of trans-1-[(2-phenylcyclopropyl)methyl]-4-arylpiperazines[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Zhang et al.
Journal of medicinal chemistry, 43(21), 3923-3932 (2000-10-29)
The dopaminergic receptor profile of a series of trans-1-[(2-phenylcyclopropyl)methyl]-4-arylpiperazines was examined. Aromatic substitution patterns were varied with the goal of identifying a compound having affinities for the D(2) and D(4) receptors in a ratio similar to that observed for the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service