Skip to Content
Merck
All Photos(1)

Key Documents

132691

Sigma-Aldrich

3-Chloropropionic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2COOH
CAS Number:
Molecular Weight:
108.52
Beilstein:
1098495
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Product 132691 is not currently sold in your country. Contact Technical Service

Quality Level

Assay

98%

form

solid

bp

203-205 °C (lit.)

mp

35-40 °C (lit.)

solubility

H2O: soluble 50 mg/mL, clear, colorless

functional group

carboxylic acid
chloro

SMILES string

OC(=O)CCCl

InChI

1S/C3H5ClO2/c4-2-1-3(5)6/h1-2H2,(H,5,6)

InChI key

QEYMMOKECZBKAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Chloropropionic acid is used in pesticides[1].

Application

3-Chloropropionic acid was used to study the degradation of 3-chloropropionic acid by Pseudomonas sp. strain isolated from rice paddy fields[1]. It was also used in determination of 3-bromopropionic acid, biomarker for the exposure to 1-bromopropane in human urine by gas chromatography[2].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Degradation of 3-chloropropionic acid (3CP) byPseudomonas sp. B6P isolated from a rice paddy field.
Mesri S, et al.
Annals of Microbiology, 59(3), 447-451 (2009)
Gang Cheng et al.
International journal of nanomedicine, 7, 3991-4006 (2012-08-21)
The reduction-sensitive cationic polymer is a promising nonviral carrier for gene delivery. Until now, disulfide bonds have been the only golden standard for its design. The aim of this research was to develop a novel reduction-responsive cationic polymer as a
Alba T Macias et al.
Bioorganic & medicinal chemistry, 12(7), 1643-1647 (2004-03-19)
Gamma-Hydroxybutyric acid (GHB) has gained in notoriety in recent years due to its association with sexual assaults. GHB is an endogenous ligand for GHB receptors, but its complete pharmacological mechanism of action in vivo remains unclear due to apparent GABAergic
C Rottleb et al.
International journal of cancer, 67(5), 724-729 (1996-09-04)
Terminal differentiation of hematopoietic cells in vivo and in vitro is almost invariably accompanied by down-regulated expression of the c-myc proto-oncogene. Constitutive expression of c-myc in tumor cells inhibits terminal differentiation and maintains proliferation. In Burkitt's lymphoma, chromosomal translocations cause
Wanwen Chen et al.
International journal of biological macromolecules, 173, 504-512 (2021-01-19)
Synthetic selenium polysaccharides with potential bioactivity have drawn great interest due to the SeO bonds existing in the structure. Herein, N, O-selenized N-(2-carboxyethyl) chitosan (sNCCS) was synthesized through carboxyethylation and selenylation. Various characterizations were performed to identify the structure of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service